Learn More
Cole-Parmer Techne FBATH26 Unheated Water Bath, Stainless Steel, 26L
Supplier:  Cole-Parmer 1610189
Encompass_Preferred
Designed to be used with a clip-on Tempette or Tempunit thermoregulator (sold separately), these water baths incorporate carrying handles for added safety. All water baths have stoved enamelled steel outer cases and are supplied with bridge mounting plate to hold the thermoregulator.
Catalog No. 50-196-0473