Learn More
SPEX CERTIPREP LLC 3P 26 QUAL CONTROL STANDARD 50ML
Catalog No. NC0750549
Encompass_Preferred
(HPS by Zeptometrix) Quality Control Std 26: Si - 50 ug/mL, Al, Sb, As, B, Ba, Be, Ca, Cd, Co, Cr, Cu, Fe, Mg, Mn, Mo, Na, Ni, Pb, Ag, Se, Ti, Tl, V, Zn - 100 ug/mL, K - 1,000 ug/mL in 4% HNO3 + Tr HF Exp Date: 1 Year
Catalog No. NC0750549 Supplier SPEX CERTIPREP LLC 3P Supplier No. QCS26100
(HPS by Zeptometrix) Quality Control Std 26: Si - 50 ug/mL, Al, Sb, As, B, Ba, Be, Ca, Cd, Co, Cr, Cu, Fe, Mg, Mn, Mo, Na, Ni, Pb, Ag, Se, Ti, Tl, V, Zn - 100 ug/mL, K - 1,000 ug/mL in 4% HNO3 + Tr HF Exp Date: 1 Year