Learn More
Hans Rudolph Inc 2600 MED TWO-WAY T-SHAPE NRBV
SDP
Catalog No. NC2120866
Encompass_Preferred
Medium Two-Way Non-Rebreathing T-Shape™ Valve. The Mouth Port Tube is 1.125”(28.6mm)OD/22mmlD taper. Inhalation andExhalation Port Tubes are 1.375”(35.0mm)OD/1.125”(28.6mm)lD
This item is not returnable. View return policy
Catalog No. NC2120866 Supplier Hans Rudolph Inc Supplier No. 112421
 Restricted Item
Add to Cart
Add to Cart
Medium Two-Way Non-Rebreathing T-Shape™ Valve. The Mouth Port Tube is 1.125”(28.6mm)OD/22mmlD taper. Inhalation andExhalation Port Tubes are 1.375”(35.0mm)OD/1.125”(28.6mm)lD