Learn More
Thermco Products Inc General lab Hg thermometer, -10 deg./260 deg.C & 20 deg./500 deg.F, 1 deg.C div & 2 deg.F, 76mm imm, 405mm
Catalog No. NC0519248
Encompass_Preferred
Mercury filled double scale thermometer, yellow back tubing, with toughened bulbs. Suspension ring for hanging, vertical numbering. Individual serial numbered. Packed in non-roll square tubing. Permanent markings.
Catalog No. NC0519248 Supplier Thermco Products Inc Supplier No. B260C500FW3
Mercury filled double scale thermometer, yellow back tubing, with toughened bulbs. Suspension ring for hanging, vertical numbering. Individual serial numbered. Packed in non-roll square tubing. Permanent markings.