Learn More
Aviva Systems Biology Corp 2610034B18Rik Antibody - N-terminal region (ARP55834_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55834_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against 2610034B18Rik. It was validated on Western Blot. . Alternate names: Arpin; C15orf38ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-286-089