Learn More
Refrigiwear Inc. 2650 Reg Gray Sml Cowhide Freezer Dexterity Glove
SDP
Catalog No. NC3885903
Encompass_Preferred
Enjoy the durability of a leather freezer glove without added bulk that limits your dexterity. The thin layers of foam and fiberfill insulation in these warm work gloves keep your fingers warm while the soft split suede cowhide leather makes it easy to keep them moving, even at a bone-chilling -20°F. Comfort Rating: -20°F 100% Polyester foam and fiberfill insulation Split suede cowhide leather Soft brushed tricot lining Pre-curved, ergonomic fit Heavy-duty knit wrist    
Catalog No. NC3885903 Supplier Refrigiwear Inc. Supplier No. 2650RGRASML
Enjoy the durability of a leather freezer glove without added bulk that limits your dexterity. The thin layers of foam and fiberfill insulation in these warm work gloves keep your fingers warm while the soft split suede cowhide leather makes it easy to keep them moving, even at a bone-chilling -20°F. Comfort Rating: -20°F 100% Polyester foam and fiberfill insulation Split suede cowhide leather Soft brushed tricot lining Pre-curved, ergonomic fit Heavy-duty knit wrist    