Learn More
Waters Corp Waters Corporation GPV Assembly, Gradient Proportioning Valve, HPLC System
Catalog No. 50818936 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement Gradient Proportioning Valve (GPV) Assembly is engineered for precise fluid management within separation modules and HPLC systems. It ensures exact, reproducible mixing of up to four different solvents through its finely toleranced opening and closing rates and aperture sizes. The assembly’s four inlet valves are solenoid-actuated, incorporating precise electrical characteristics to mitigate heat buildup and safeguard against outgassing.
  • Features precise opening and closing rates and aperture sizes with exceptionally tight tolerance.
  • Four inlet valves are actuated by solenoids that have precise electrical characteristics.
  • Helps reduce heat buildup and protect the system from outgassing.
  • Provides exact, reproducible mixing of up to four different solvents.
  • Used in separation modules.
  • Component for high performance liquid chromatography systems.
Catalog No. 50-818-936 Supplier Waters Corp Supplier No. WAT270927
This replacement Gradient Proportioning Valve (GPV) Assembly is engineered for precise fluid management within separation modules and HPLC systems. It ensures exact, reproducible mixing of up to four different solvents through its finely toleranced opening and closing rates and aperture sizes. The assembly’s four inlet valves are solenoid-actuated, incorporating precise electrical characteristics to mitigate heat buildup and safeguard against outgassing.
  • Features precise opening and closing rates and aperture sizes with exceptionally tight tolerance.
  • Four inlet valves are actuated by solenoids that have precise electrical characteristics.
  • Helps reduce heat buildup and protect the system from outgassing.
  • Provides exact, reproducible mixing of up to four different solvents.
  • Used in separation modules.
  • Component for high performance liquid chromatography systems.