Learn More
Waters Corp Waters Performance Maintenance Kit, Alliance HPLC System, 2690/95 Separations Module, Annual
Catalog No. 50819002 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Performance Maintenance Kit for the 2695 Separations Module includes all the necessary parts to keep the instrument operating at peak performance on an annual basis.
  • Diffuser assembly
  • Filter insert assembly
  • Four stainless steel solvent filters
  • Four face seals
  • Two fluoroloy seals
  • One 250μL syringe
  • One compression screw
  • One sealwash injector tube
  • One needlewash frit
  • Five sealwash tubes
  • Two seals
  • One spacer
  • Two plungers
  • One teflon washer
  • Two plunger seal wash
  • One needle wash filter
  • One filter retainer
  • Thin walled teflon tubing
  • Two check valve cartridge assemblies
  • Powder-free gloves
  • One injector needle
  • One injector needle ferrule
Catalog No. 50-819-002 Supplier Waters Corp Supplier No. WAT270944
This Performance Maintenance Kit for the 2695 Separations Module includes all the necessary parts to keep the instrument operating at peak performance on an annual basis.
  • Diffuser assembly
  • Filter insert assembly
  • Four stainless steel solvent filters
  • Four face seals
  • Two fluoroloy seals
  • One 250μL syringe
  • One compression screw
  • One sealwash injector tube
  • One needlewash frit
  • Five sealwash tubes
  • Two seals
  • One spacer
  • Two plungers
  • One teflon washer
  • Two plunger seal wash
  • One needle wash filter
  • One filter retainer
  • Thin walled teflon tubing
  • Two check valve cartridge assemblies
  • Powder-free gloves
  • One injector needle
  • One injector needle ferrule