Learn More
Waters Corp Waters Pressure Transducer, Replacement Strain Gauge, HPLC, Purification
Catalog No. 50818942 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement strain gauge transducer assembly is an advanced and reliable pressure-detection device. It offers enhanced accuracy and linearity compared to the Bourdon tube transducer and does not contain light bulbs or moving parts that could fail. This strain gauge transducer is compatible with the 515, 1515, and 1525 HPLC Pumps, as well as Alliance Separation Modules.
  • Replacement strain gauge transducer assembly.
  • Advanced and reliable pressure-detection device.
  • Offers enhanced accuracy and linearity compared to Bourdon tube transducers.
  • Contains no light bulbs or moving parts, reducing failure points.
  • Used in 515, 1515, 1525 HPLC pumps and Alliance separation modules.
Catalog No. 50-818-942 Supplier Waters Corp Supplier No. WAT270966
This replacement strain gauge transducer assembly is an advanced and reliable pressure-detection device. It offers enhanced accuracy and linearity compared to the Bourdon tube transducer and does not contain light bulbs or moving parts that could fail. This strain gauge transducer is compatible with the 515, 1515, and 1525 HPLC Pumps, as well as Alliance Separation Modules.
  • Replacement strain gauge transducer assembly.
  • Advanced and reliable pressure-detection device.
  • Offers enhanced accuracy and linearity compared to Bourdon tube transducers.
  • Contains no light bulbs or moving parts, reducing failure points.
  • Used in 515, 1515, 1525 HPLC pumps and Alliance separation modules.