Learn More
Waters Corp Seal Pack Assembly, Replacement, HPLC System, 2695 Compatible
Catalog No. 50818941 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement seal pack assembly is designed for use with HPLC systems. It features a one-piece seal pack with a brazed union for the column inlet line, ensuring reliable performance in demanding laboratory environments. This pre-assembled unit is compatible with various injector assemblies, offering broad applicability.
  • One-piece seal pack with brazed union for the column inlet line.
  • Compatible with injector assemblies, including pre-Phase 1, Phase 1, and Phase 2 designs.
  • Can be rebuilt using a separate seal pack rebuild kit.
  • Designed for HPLC system applications.
  • Replacement is scheduled as required.
Catalog No. 50-818-941 Supplier Waters Corp Supplier No. 700002791
This replacement seal pack assembly is designed for use with HPLC systems. It features a one-piece seal pack with a brazed union for the column inlet line, ensuring reliable performance in demanding laboratory environments. This pre-assembled unit is compatible with various injector assemblies, offering broad applicability.
  • One-piece seal pack with brazed union for the column inlet line.
  • Compatible with injector assemblies, including pre-Phase 1, Phase 1, and Phase 2 designs.
  • Can be rebuilt using a separate seal pack rebuild kit.
  • Designed for HPLC system applications.
  • Replacement is scheduled as required.