Learn More
Waters Corp Waters Valve, 2690 Sealwash Pump, HPLC, UPLC, Purification
Catalog No. 50818935 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This valve serves as a replacement seal wash pump solenoid, designed to facilitate the flow of seal wash solvent through pump heads. It ensures efficient operation across a range of separation modules and solvent managers in various chromatography systems.
  • Functions as a replacement seal wash pump solenoid.
  • Drives seal wash solvent through pump heads.
  • Applicable for HPLC, UPLC, and purification system types.
  • Compatible with various separation modules, solvent managers, and 25x5 pumps.
  • Recommended for replacement as required.
  • Includes one unit per package.
Catalog No. 50-818-935 Supplier Waters Corp Supplier No. WAT270926
This valve serves as a replacement seal wash pump solenoid, designed to facilitate the flow of seal wash solvent through pump heads. It ensures efficient operation across a range of separation modules and solvent managers in various chromatography systems.
  • Functions as a replacement seal wash pump solenoid.
  • Drives seal wash solvent through pump heads.
  • Applicable for HPLC, UPLC, and purification system types.
  • Compatible with various separation modules, solvent managers, and 25x5 pumps.
  • Recommended for replacement as required.
  • Includes one unit per package.