Learn More
Waters Corp Waters Solvent Inlet Tubing, Replacement Set, Alliance e2695, HPLC System
Catalog No. 50818946 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product is a replacement set of four solvent inlet lines designed for use with the Alliance e2695 HPLC System. It includes all necessary components for installation.
  • Designed for Alliance e2695 Separations Module
  • Suitable for HPLC systems
  • Includes four solvent inlet lines (A-D)
  • Contains a solvent filter/sinker
  • Supplied with a label and connector to the degasser
  • Provided as a single package
Catalog No. 50-818-946 Supplier Waters Corp Supplier No. WAT271022
This product is a replacement set of four solvent inlet lines designed for use with the Alliance e2695 HPLC System. It includes all necessary components for installation.
  • Designed for Alliance e2695 Separations Module
  • Suitable for HPLC systems
  • Includes four solvent inlet lines (A-D)
  • Contains a solvent filter/sinker
  • Supplied with a label and connector to the degasser
  • Provided as a single package