Learn More
World Precision Instrument 26g Beveled Replacement NanoFil needles
SDP
Catalog No. 502521063
Encompass
Our beveled tips are shorter, more durable, and offer better penetration than standard removable needles.
Catalog No. 50-252-1063 Supplier World Precision Instrument Supplier No. NF26BV2
Our beveled tips are shorter, more durable, and offer better penetration than standard removable needles.