Learn More
Jensen Global Inc 26G,DISP TIP,PEACH,STRGHT,50/P
Catalog No. NC2232242
Encompass_Preferred
26 GAUGE NT PREMIUM SERIES DISPENSING TIPPeach colored Luer Lock hub with a 26 gauge blunt tip. The Inside Diameter is 0.009” (0.240 mm) and the Outside Diameter is 0.018” (0.450 mm). Our NT Series Premium polished tips feature a polypropylene plastic double-helix hub with a UV-bonded stainless steel cannula. The burr-free tip design allows for precise dispensing with no residuals. Available in boxes of 50 or bags of 1000.Gauge: 26 Color: PeachID: 0.009" (0.240 mm)OD: 0.018" (0.450 mm)
This item is not returnable. View return policy
Catalog No. NC2232242 Supplier Jensen Global Inc Supplier No. JG260.5HPX
26 GAUGE NT PREMIUM SERIES DISPENSING TIPPeach colored Luer Lock hub with a 26 gauge blunt tip. The Inside Diameter is 0.009” (0.240 mm) and the Outside Diameter is 0.018” (0.450 mm). Our NT Series Premium polished tips feature a polypropylene plastic double-helix hub with a UV-bonded stainless steel cannula. The burr-free tip design allows for precise dispensing with no residuals. Available in boxes of 50 or bags of 1000.Gauge: 26 Color: PeachID: 0.009" (0.240 mm)OD: 0.018" (0.450 mm)