Learn More
Laddawn 26x36 4mil, Layflat Bags, No Color Additives, 200/Case
Catalog No. NC1468437
Encompass
26x36 4mil, Bags, 200/case.Contains no color additives. Made with a proprietary blend of 100% virgin low and linear low density polyethylene. Provided in cases.
Catalog No. NC1468437 Supplier Laddawn Supplier No. 1289
26x36 4mil, Bags, 200/case.Contains no color additives. Made with a proprietary blend of 100% virgin low and linear low density polyethylene. Provided in cases.