Learn More
Cayman Chemical 27-alkyn Cholesterol 100ug
SDP
Catalog No. 502936509
Encompass_Preferred
An alkyne derivative of cholesterol for click chemistry; has been used to track cholesterol metabolism and localization
Catalog No. 50-293-6509 Supplier Cayman Chemical Supplier No. 24319100 UG
An alkyne derivative of cholesterol for click chemistry; has been used to track cholesterol metabolism and localization