Learn More
Medex Supply 27 GAUGE NEEDLE 3/4IN 50/PK
SDP
Catalog No. NC9063112
Encompass_Preferred
Exel Scalp Vein Butterfly Sets, 27G x 3/4", Gray, 50/pk. Thin wall stainless steel cannula, siliconized for easy insertion. Flexible wings hold the needle in position and help to prevent inadvertent pull out. Sterilized with Ethylene Oxide and ready for immediate use. Color-coded for easier identification. Large diameter needle permits greater flow of fluids. 12" tube length.
This item is not returnable. View return policy
Catalog No. NC9063112 Supplier Medex Supply Supplier No. EXL26709
Exel Scalp Vein Butterfly Sets, 27G x 3/4", Gray, 50/pk. Thin wall stainless steel cannula, siliconized for easy insertion. Flexible wings hold the needle in position and help to prevent inadvertent pull out. Sterilized with Ethylene Oxide and ready for immediate use. Color-coded for easier identification. Large diameter needle permits greater flow of fluids. 12" tube length.