Learn More
Medchemexpress LLC HY-N2371 1mg Medchemexpress, 27-Hydroxycholesterol CAS:20380-11-4 Purity:>98%
SDP
Catalog No. 501933923
Encompass_Preferred
Medchemexpress, HY-N2371 1mg 27-Hydroxycholesterol CAS:20380-11-4 27-Hydroxycholesterol is a selective estrogen receptor modulator and an agonist of the liver X receptor . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3923 Supplier Medchemexpress LLC Supplier No. HYN23711MG
Medchemexpress, HY-N2371 1mg 27-Hydroxycholesterol CAS:20380-11-4 27-Hydroxycholesterol is a selective estrogen receptor modulator and an agonist of the liver X receptor . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.