Learn More
Enzo Life Sciences 27-Hydroxycholesterol (1mg). CAS: 20380-11-4
SDP
Catalog No. 502013376
Encompass_Preferred
27-Hydroxycholesterol (27-HC) is a metabolite resulting from the action of sterol 27-Hydroxylase on cholesterol. It is an endogenous selective estrogen receptor modulator and ligand for the liver X receptor (LXR). 27-Hydroxycholesterol plays a role in Alzheimer's disease, oxidative stress, metabolism, cell-signaling, and heart disease. Alternative name: 5,25R-​Cholest​en-​3β,​26-​diol, (25R)-26-Hydroxycholesterol. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (up to 10 mg/ml with warming) or in ethanol (up to 10 mg/ml). Insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-3376 Supplier Enzo Life Sciences Supplier No. ENZCHM1020001
27-Hydroxycholesterol (27-HC) is a metabolite resulting from the action of sterol 27-Hydroxylase on cholesterol. It is an endogenous selective estrogen receptor modulator and ligand for the liver X receptor (LXR). 27-Hydroxycholesterol plays a role in Alzheimer's disease, oxidative stress, metabolism, cell-signaling, and heart disease. Alternative name: 5,25R-​Cholest​en-​3β,​26-​diol, (25R)-26-Hydroxycholesterol. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (up to 10 mg/ml with warming) or in ethanol (up to 10 mg/ml). Insoluble in water. Long Term Storage: -20°C.