Learn More
Medchemexpress LLC 27-Norcholest-5-en-25-yn-3-ol, (3β)- | 1631985-09-5 | C26H40O | 5 MG
SDP
Catalog No. 5000372426
Encompass_Preferred
Alkyne Cholesterol, a modified lipid, is a click chemistry reagent containing an alkyne group. The terminal alkyne group can be used in a highly specific linking reaction with azide-containing reagents in the presence of a copper (Cu)-containing catalyst. Alkyne Cholesterol can be used for tracking cellular cholesterol metabolism and localization.
  • It is a click chemistry reagent containing an alkyne group.
  • The terminal alkyne group can be used in a highly specific linking reaction with azide-containing reagents in the presence of a copper (Cu)-containing catalyst.
  • It can be used for tracking cellular cholesterol metabolism and localization.
Catalog No. 50-003-72426 Supplier Medchemexpress LLC Supplier No. HY1517055MG
Alkyne Cholesterol, a modified lipid, is a click chemistry reagent containing an alkyne group. The terminal alkyne group can be used in a highly specific linking reaction with azide-containing reagents in the presence of a copper (Cu)-containing catalyst. Alkyne Cholesterol can be used for tracking cellular cholesterol metabolism and localization.
  • It is a click chemistry reagent containing an alkyne group.
  • The terminal alkyne group can be used in a highly specific linking reaction with azide-containing reagents in the presence of a copper (Cu)-containing catalyst.
  • It can be used for tracking cellular cholesterol metabolism and localization.