Learn More
Gilson Company Inc 270 ALPINE W/SEAL
Catalog No. NC9203644
Encompass_Preferred
Micron Air Jet Sieve, Size: no.270 (53micrometer), Standard 200mm dia., Stainless Steel Frame and Mesh
Catalog No. NC9203644 Supplier Gilson Company Inc Supplier No. AJA216
Micron Air Jet Sieve, Size: no.270 (53micrometer), Standard 200mm dia., Stainless Steel Frame and Mesh