Learn More
Waters Corp Performance Maintenance Kit, 2707 Autosampler (Preparative), HPLC, Annual
Catalog No. 502390510 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters™ Performance Maintenance Kit for the 2707 Autosampler (Preparative) contains all the Waters Quality Parts™ necessary to keep the 2707 Autosampler (Preparative) operating at peak performance on an annual basis.
  • Kit includes an air needle (50 mm).
  • Includes a 60μL needle assembly.
  • Comes with a wash station with screws.
  • Includes a syringe valve.
  • Contains buffer tubing (2000μL).
  • Includes a Prep rotor seal.
  • Comes with a 2500μL syringe.
  • Designed for annual replacement.
  • Suitable for HPLC systems.
  • Product type: Performance Maintenance Kit.
Catalog No. 50-239-0510 Supplier Waters Corp Supplier No. 201000306
The Waters™ Performance Maintenance Kit for the 2707 Autosampler (Preparative) contains all the Waters Quality Parts™ necessary to keep the 2707 Autosampler (Preparative) operating at peak performance on an annual basis.
  • Kit includes an air needle (50 mm).
  • Includes a 60μL needle assembly.
  • Comes with a wash station with screws.
  • Includes a syringe valve.
  • Contains buffer tubing (2000μL).
  • Includes a Prep rotor seal.
  • Comes with a 2500μL syringe.
  • Designed for annual replacement.
  • Suitable for HPLC systems.
  • Product type: Performance Maintenance Kit.