Learn More
Waters Corp 2747 Performance Maintenance Kit
Supplier:  Waters Corp 201000132
Encompass_Preferred
The Waters Performance Maintenance Kit for the 2747 sample manager contains all the Waters Quality Parts necessary to keep your 2747 sample manager operating at peak performance on an annual basis. Waters Performance Maintenance Kit contains the following parts: 1.0mL syringe, 2767/2747 Tubing Kit, vented probe, lubrication kit, injector port fittings kit, 3-port valve and Rheodyne analytical injector rebuild kit.
Catalog No. 50-791-555