Learn More
Waters Corp Waters Performance Maintenance Kit, 2767 Sample Manager, HPLC, Annual
Catalog No. 50791584 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Performance Maintenance Kit is designed for the 2767 sample manager, containing essential components to maintain instrument peak performance on an annual basis. It includes a variety of quality parts necessary for optimal operation, ensuring your system continues to function reliably.
  • Vented probe included
  • Lubrication kit included
  • Injector port fittings kit included
  • Rheodyne analytical injector rebuild kit included
  • Rheodyne prep injector rebuild kit included
  • Designed for HPLC systems
  • Recommended for annual replacement
  • Syringes are recommended for annual replacement and purchased separately as they are customer configurable
  • One unit per package
Catalog No. 50-791-584 Supplier Waters Corp Supplier No. 201000195
This Performance Maintenance Kit is designed for the 2767 sample manager, containing essential components to maintain instrument peak performance on an annual basis. It includes a variety of quality parts necessary for optimal operation, ensuring your system continues to function reliably.
  • Vented probe included
  • Lubrication kit included
  • Injector port fittings kit included
  • Rheodyne analytical injector rebuild kit included
  • Rheodyne prep injector rebuild kit included
  • Designed for HPLC systems
  • Recommended for annual replacement
  • Syringes are recommended for annual replacement and purchased separately as they are customer configurable
  • One unit per package