Learn More
Contec™ 2771 Bucket System
BIOPHARMA
Catalog No. 19159275 Shop All Contec Inc. Products
This item is not returnable. View return policy
Catalog No. 19-159-275 Supplier Contec™ Supplier No. 2771KIT
This item is not returnable. View return policy
Includes
Two polypropylene buckets (gray, red), stainless steel drip cap
Ideal for use in smaller cleanrooms.
  • Fully autoclavable
  • Stainless steel rolling cart with standard casters
  • Easy to maneuver around any cleanroom
  • Allows for differentiation between cleaning and rinsing solutions