Learn More
Waters Corp Waters Sample Loop, 50 μL, for 2790/2795 Separations Module
Catalog No. 50792409 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 2790 Separations Module utilizes a standard 50μL sample loop. This loop facilitates injections of 50μL samples when operating in Full Loop mode, and injections of samples less than 50μL in partial loop mode.
  • Designed for use with the 2790 Separations Module
  • Volume of 50 μL
  • Supports 50 μL sample injections in Full Loop mode
  • Supports sample injections less than 50 μL in partial loop mode
  • As required replacement schedule
Catalog No. 50-792-409 Supplier Waters Corp Supplier No. 700000560
The 2790 Separations Module utilizes a standard 50μL sample loop. This loop facilitates injections of 50μL samples when operating in Full Loop mode, and injections of samples less than 50μL in partial loop mode.
  • Designed for use with the 2790 Separations Module
  • Volume of 50 μL
  • Supports 50 μL sample injections in Full Loop mode
  • Supports sample injections less than 50 μL in partial loop mode
  • As required replacement schedule