Learn More
Waters Corp Waters Injector Valve PM Kit, 2790/95 System, Rotor Seal, Stator Face Assembly
Catalog No. 50818992 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters Performance Maintenance Kit is designed to ensure the optimal operation of injector valves. It contains all the necessary quality parts to maintain peak performance in compatible systems.
  • Contains all necessary parts to maintain peak performance
  • Includes rotor seal
  • Includes stator face assembly
  • Designed for 2790/95 injector valve systems
  • Type: Performance Maintenance Kit
  • System type: HPLC
  • Replacement schedule: As Required
Catalog No. 50-818-992 Supplier Waters Corp Supplier No. 201000124
The Waters Performance Maintenance Kit is designed to ensure the optimal operation of injector valves. It contains all the necessary quality parts to maintain peak performance in compatible systems.
  • Contains all necessary parts to maintain peak performance
  • Includes rotor seal
  • Includes stator face assembly
  • Designed for 2790/95 injector valve systems
  • Type: Performance Maintenance Kit
  • System type: HPLC
  • Replacement schedule: As Required