Learn More
Waters Corp Waters Needle, Stainless Steel, for 2790 and 2795 Systems
Catalog No. 50818907 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product is a stainless steel needle for chromatography systems. It is designed for use with 2790 and 2795 systems, offering reliable performance for various laboratory applications.
  • Made from stainless steel
  • Prevents coring or needle degradation
  • Compatible with pre-slit septa, sealing foils, or film
  • Resolves “needle bottom sensor error” caused by bent needles
  • Ensures proper system initialization and operation
Catalog No. 50-818-907 Supplier Waters Corp Supplier No. 700000389
This product is a stainless steel needle for chromatography systems. It is designed for use with 2790 and 2795 systems, offering reliable performance for various laboratory applications.
  • Made from stainless steel
  • Prevents coring or needle degradation
  • Compatible with pre-slit septa, sealing foils, or film
  • Resolves “needle bottom sensor error” caused by bent needles
  • Ensures proper system initialization and operation