Learn More
Waters Corp Waters Sample Loop, 500 μL
Catalog No. 50818910 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The optional 500μL sample loop is one of the larger sample loops which can be installed to inject full loop sample volumes other than 50 μL.
  • Injects full loop sample volumes other than 50 μL.
  • Provides a volume of 500 μL.
  • Supplied as 1 unit per package.
Catalog No. 50-818-910 Supplier Waters Corp Supplier No. 700000562
The optional 500μL sample loop is one of the larger sample loops which can be installed to inject full loop sample volumes other than 50 μL.
  • Injects full loop sample volumes other than 50 μL.
  • Provides a volume of 500 μL.
  • Supplied as 1 unit per package.