Learn More
Waters Corp Waters Corporation Washer, Inject Port Seal, for 2790/2795 Separations Module
Catalog No. 50818906 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Inject Port Seal Washer is located in the inject port of the 2790/2795 Separations Module. It requires replacement if it is leaking or obstructing the inject port flowpath. This component is replaced simultaneously with the inject port seal washer and the inject port seat.
  • Ensures proper sealing within the inject port.
  • Prevents leaks in the inject port.
  • Maintains clear flowpath within the inject port.
Catalog No. 50-818-906 Supplier Waters Corp Supplier No. 700000385
The Inject Port Seal Washer is located in the inject port of the 2790/2795 Separations Module. It requires replacement if it is leaking or obstructing the inject port flowpath. This component is replaced simultaneously with the inject port seal washer and the inject port seat.
  • Ensures proper sealing within the inject port.
  • Prevents leaks in the inject port.
  • Maintains clear flowpath within the inject port.