Learn More
Waters Corp Waters Inject Port Seat, For 2790/2795 Separations Module
Catalog No. 50819180 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Inject Port Seat is located in the inject port of the 2790/2795 Separations Module. It establishes a fluid connection from the needle to the sample loop.
  • Establishes a fluid connection from the needle to the sample loop.
  • Designed for use in 2790/2795 Separations Modules.
  • Engineering code: 415000181.
Catalog No. 50-819-180 Supplier Waters Corp Supplier No. 700000384
The Inject Port Seat is located in the inject port of the 2790/2795 Separations Module. It establishes a fluid connection from the needle to the sample loop.
  • Establishes a fluid connection from the needle to the sample loop.
  • Designed for use in 2790/2795 Separations Modules.
  • Engineering code: 415000181.