Learn More
Waters Corp Syringe, 500 µL, 2795, Standard
Catalog No. 50818911 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 2790 Separations Module is equipped with a standard 500µL syringe. This syringe, coupled with the standard 50µL sample loop and 128µL holding loop provides good accuracy and repeatability performance in a short amount of time for typical chromatographic sample volumes (5 to 50µL).
Catalog No. 50-818-911 Supplier Waters Corp Supplier No. 700000565
The 2790 Separations Module is equipped with a standard 500µL syringe. This syringe, coupled with the standard 50µL sample loop and 128µL holding loop provides good accuracy and repeatability performance in a short amount of time for typical chromatographic sample volumes (5 to 50µL).