Learn More
Waters Corp Sample Loop, 5 μL, PEEK
Supplier: Waters Corp 430000781
The Waters Sample Loop, 5 μL, PEEK, is a crucial component of the sample management system within the Waters e2796 Bioseparations Module. It plays a vital role in precise sample introduction by managing the flow path of samples.
- Part of the sample management system
- Connects to the inject valve
- Isolates sample loop from solvent flow path to the column during loading
- Connects sample loop to solvent flow during injection
- Facilitates precise sample introduction