Learn More
SAI Infusion Technologies 27 Gauge, 0.5" Butterfly Needle - 24cm PU Tubing
SDP
Catalog No. NC2243844
Encompass
27G0.5" Butterfly (Winged Infusion Set) with 24cm PU tubing. For access to the tubing, use a 27G blunt needle such as the B27-50. Product is for laboratory animal research only.
This item is not returnable. View return policy
Catalog No. NC2243844 Supplier SAI Infusion Technologies Supplier No. BFL24
27G0.5" Butterfly (Winged Infusion Set) with 24cm PU tubing. For access to the tubing, use a 27G blunt needle such as the B27-50. Product is for laboratory animal research only.