Learn More
SAI Infusion Technologies 27 Gauge, 0.5" Butterfly Needle - 5cm PU Tubing
SDP
Catalog No. NC3355225
Encompass
27G0.5" Butterfly (Winged Infusion Set) with 5cm PU tubing. For access to the tubing, use a 27G blunt needle such as the B27-50. Product is for laboratory animal research only.
Catalog No. NC3355225 Supplier SAI Infusion Technologies Supplier No. BF2713
27G0.5" Butterfly (Winged Infusion Set) with 5cm PU tubing. For access to the tubing, use a 27G blunt needle such as the B27-50. Product is for laboratory animal research only.