Learn More
SAI Infusion Technologies 12" Sampling Line with 27G, 0.5"Needle and Female Luer
SDP
Catalog No. NC3386315
Encompass
12" PU Sampling Line with an integrated 27G 0.5" hypodermic needle and a luer for easy attachment to a syringe. Use this line for dosing or for sampling. Product is for laboratory animal research only.
Catalog No. NC3386315 Supplier SAI Infusion Technologies Supplier No. BF2712C
12" PU Sampling Line with an integrated 27G 0.5" hypodermic needle and a luer for easy attachment to a syringe. Use this line for dosing or for sampling. Product is for laboratory animal research only.