Learn More
Thermo Scientific Cellulose filters, 27 mm, 100pk
Supplier:  Thermo Scientific 068093
Encompass
Diameter 27mm; Type D28; Cellulose ASE; for 1, 5, 10 or 22 mL cells; 100/PK
Catalog No. NC9048955