Learn More
Ace Glass, Inc. 27MM GRADED SEALS 27MM GRADED SEALS
SDP
Catalog No. 50974603
Encompass
27mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.
Catalog No. 50-974-603 Supplier Ace Glass, Inc. Supplier No. 845538
27mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.