Learn More
Ace Glass, Inc. 28/11 BALL JOINT (GLASSBLOWER)
SDP
Catalog No. 50975270
Encompass_Preferred
Full length, standard wall 28/11 ball joint. This ACE ball joint is precision ground and vacuum leak-tested for fit and interchangeability.
Catalog No. 50-975-270 Supplier Ace Glass, Inc. Supplier No. 765528
Full length, standard wall 28/11 ball joint. This ACE ball joint is precision ground and vacuum leak-tested for fit and interchangeability.