Learn More
Chemglass Life Sciences Joint, 28/12, Socket, Clamp Size 28, 16mm Tubing
SDP
Catalog No. CG12107
Encompass_Preferred
Joint, 28/12, Socket, Clamp Size 28, 16mm Tubing. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.
Catalog No. CG12107 Supplier Chemglass Life Sciences Supplier No. CG12107
Joint, 28/12, Socket, Clamp Size 28, 16mm Tubing. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.