Learn More
Ace Glass, Inc. 28/12 socket joint, stainless steel, unthreaded
SDP
Catalog No. 50979420
Encompass
28/12, unthreaded, socket joint fabricated from 18-8 type 303 stainless steel. Fully interchangeable with ACE glass joints and has the same I.D.
Catalog No. 50-979-420 Supplier Ace Glass, Inc. Supplier No. 765834
28/12, unthreaded, socket joint fabricated from 18-8 type 303 stainless steel. Fully interchangeable with ACE glass joints and has the same I.D.