Learn More
Ace Glass, Inc. 28/15 ball joint, stainless steel, unthreaded
SDP
Catalog No. 50975297
Encompass
28/15, unthreaded, ball joint fabricated from 18-8 type 303 stainless steel. Fully interchangeable with ACE glass joints and has the same I.D.
Catalog No. 50-975-297 Supplier Ace Glass, Inc. Supplier No. 765838
28/15, unthreaded, ball joint fabricated from 18-8 type 303 stainless steel. Fully interchangeable with ACE glass joints and has the same I.D.