Learn More
Chemglass Life Sciences Joint, 28/15, Polished Socket, Clamp Size 28, 18.80-19.20mm Tubing
SDP
Catalog No. CG121P04
Encompass_Preferred
Joint, 28/15, Polished Socket, Clamp Size 28, 18.80-19.20mm Tubing. For use with CG-122, o-ring ball joint to provide for a flexible high vacuum seal. Polished sealing surface does not abrade o-ring. Note: Not for use with CG-120 ground ball joints.
Catalog No. CG121P04 Supplier Chemglass Life Sciences Supplier No. CG121P04
Joint, 28/15, Polished Socket, Clamp Size 28, 18.80-19.20mm Tubing. For use with CG-122, o-ring ball joint to provide for a flexible high vacuum seal. Polished sealing surface does not abrade o-ring. Note: Not for use with CG-120 ground ball joints.