Learn More
Ace Glass, Inc. Adapter, 28/15 socket joint, 10mm tube end
SDP
Catalog No. 50971773
Encompass_Preferred
28/15 socket joint adapter with 10mm tube end, for joining jacketed reactor ball joint inlet/outlets to recirculator hoses.
Catalog No. 50-971-773 Supplier Ace Glass, Inc. Supplier No. 522105
28/15 socket joint adapter with 10mm tube end, for joining jacketed reactor ball joint inlet/outlets to recirculator hoses.