Learn More
Ace Glass, Inc. Adapter, 28/15 socket joint, #11 Ace-Thred end
SDP
Catalog No. 50971774
Encompass_Preferred
28/15 socket joint adapter with #11 Ace-Thred end, for joining jacketed reactor ball joint inlet/outlets to recirculator hoses.
Catalog No. 50-971-774 Supplier Ace Glass, Inc. Supplier No. 522109
28/15 socket joint adapter with #11 Ace-Thred end, for joining jacketed reactor ball joint inlet/outlets to recirculator hoses.