Learn More
Chemglass Life Sciences 28/15 Socket Joint, 19.0mm OD Tube, Clamp Size = 28
SDP
Catalog No. 501531913
Encompass_Preferred
28/15 Socket Joint, 19.0mm OD Tube, Clamp Size = 28. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.
Catalog No. 50-153-1913 Supplier Chemglass Life Sciences Supplier No. CGB12108
28/15 Socket Joint, 19.0mm OD Tube, Clamp Size = 28. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.