Learn More
Ace Glass, Inc. 28/15 socket joint, stainless steel, unthreaded
SDP
Catalog No. 50975298
Encompass
28/15, unthreaded, socket joint fabricated from 18-8 type 303 stainless steel. Fully interchangeable with ACE glass joints and has the same I.D.
Catalog No. 50-975-298 Supplier Ace Glass, Inc. Supplier No. 765840
28/15, unthreaded, socket joint fabricated from 18-8 type 303 stainless steel. Fully interchangeable with ACE glass joints and has the same I.D.