Learn More
Ace Glass, Inc. 28/15 socket joint, stainless steel, 1/2" male NPT
SDP
Catalog No. 50979424
Encompass
28/15 socket joint, fabricated from 18-8 type 303 stainless steel, is fully interchangeable with ACE glass joints, and has same size I.D. Shank is machined to 1/2 male NPT.
Catalog No. 50-979-424 Supplier Ace Glass, Inc. Supplier No. 7658240
28/15 socket joint, fabricated from 18-8 type 303 stainless steel, is fully interchangeable with ACE glass joints, and has same size I.D. Shank is machined to 1/2 male NPT.