Learn More
Chemglass Life Sciences Stopper, Spherical, Cap Type, Size 28/15
SDP
Catalog No. CG301403
Encompass_Preferred
Stopper, Spherical, Cap Type, Size 28/15. Cap style stopper to be used with apparatus having a spherical ball joint. For clamp see CG-150.
Catalog No. CG301403 Supplier Chemglass Life Sciences Supplier No. CG301403
Stopper, Spherical, Cap Type, Size 28/15. Cap style stopper to be used with apparatus having a spherical ball joint. For clamp see CG-150.