Learn More
Chemglass Life Sciences 28-400 G.P.I. Glass Thread, 25mm OD Tubing, 125mm OAL
SDP
Catalog No. CG19010
Encompass_Preferred
28-400 G.P.I. Glass Thread, 25mm OD Tubing, 125mm OAL. Thread for replacement on apparatus using G.P.I. (Glass Packaging Institute) thread.PLEASE NOTE: GPI THREADS ARE SOLD IN PACKAGES ONLY.
Catalog No. CG19010 Supplier Chemglass Life Sciences Supplier No. CG19010
28-400 G.P.I. Glass Thread, 25mm OD Tubing, 125mm OAL. Thread for replacement on apparatus using G.P.I. (Glass Packaging Institute) thread.PLEASE NOTE: GPI THREADS ARE SOLD IN PACKAGES ONLY.